missing translation for 'onlineSavingsMsg'
Learn More

1,7,7-Trimethylbicyclo[2.2.1]heptan-2-amine hydrochloride, 97%, Thermo Scientific™

Product Code. 10217762
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10217762 1 g 1g
10066764 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10217762 Supplier Thermo Scientific Maybridge Supplier No. RDP00128DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 32768-19-7
Molecular Formula C10H20ClN
Molecular Weight (g/mol) 189.727
MDL Number MFCD00180693
InChI Key XVVITZVHMWAHIG-VMKQZJJLSA-N
Synonym 1,7,7-trimethylbicyclo 2.2.1 heptan-2-amine hydrochloride, bornylamine, hydrochloride, bornylamine hydrochloride, 1,7,7-trimethylnorbornan-2-amine, 1,7,7-trimethylbicyclo 2.2.1 hept-2-ylamine, chloride, 4,7,7-trimethylbicyclo 2.2.1 heptan-3-amine hydrochloride
PubChem CID 55283045
IUPAC Name (1R,4R)-4,7,7-trimethylbicyclo[2.2.1]heptan-3-amine;hydrochloride
SMILES CC1(C2CCC1(C(C2)N)C)C.Cl

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 189.73
Percent Purity 97%
Chemical Name or Material 1, 7, 7-Trimethylbicyclo[2.2.1]heptan-2-amine hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.