missing translation for 'onlineSavingsMsg'
Learn More

1,6-Naphthyridine, 96%, Thermo Scientific™

Product Code. 15535986 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15535986 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15535986 Supplier Thermo Scientific Acros Supplier No. 457670010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 253-72-5
Melting Point 36.0°C to 38.0°C
Infrared Spectrum Authentic
Assay Percent Range 95% min. (GC)
Packaging Glass Bottle
Molecular Formula C8H6N2
MDL Number MFCD00059750
Quantity 1 g
Synonym 1,6-diazanaphthalene, 1,6 naphthyridine, 1,6-pyridopyridine, unii-s1t14p7ow7, pyridino 4,3-b pyridine, 1,6naphthyridine, 1,6-naphthyridine, pubchem18071, acmc-1chvm, pyrido 4,3-b pyridine
InChI Key VSOSXKMEQPYESP-UHFFFAOYSA-N
SMILES C1=CN=C2C=CN=CC2=C1
IUPAC Name 1,6-naphthyridine
Molecular Weight (g/mol) 130.15
PubChem CID 67488
ChEBI CHEBI:36627
Formula Weight 130.15
Percent Purity 96%
Chemical Name or Material 1,6-Naphthyridine
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.