missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Methylpyrazin-2-yl)piperidine-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10515374
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10515374 1 g Amber Glass Bottle 1g
10317304 250 mg Amber Glass Bottle 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10515374 Supplier Thermo Scientific Maybridge Supplier No. CC59001DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 930111-02-7
Molecular Formula C11H15N3O2
Molecular Weight (g/mol) 221.26
MDL Number MFCD09879914
InChI Key TTWSTFTZNIMXTA-UHFFFAOYSA-N
Synonym 1-6-methylpyrazin-2-yl piperidine-3-carboxylic acid
PubChem CID 24229596
IUPAC Name 1-(6-methylpyrazin-2-yl)piperidine-3-carboxylic acid
SMILES CC1=CN=CC(=N1)N2CCCC(C2)C(=O)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 221.26
Percent Purity 97%
Chemical Name or Material 1-(6-Methylpyrazin-2-yl)piperidine-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.