missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Methoxy-2-naphthyl)ethanol, 98%

Product Code. 11476430
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476430 5 g 5g
11486430 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11476430 Supplier Thermo Scientific Alfa Aesar Supplier No. B25628.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 77301-42-9 | C13H14O2 | 202.25 g/mol

Racemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Racemic 1-(6-methoxy-2-naphthyl)ethanol converts to the methyl ester of naproxen.

Solubility
Soluble in DMSO, acetone, dichloromethane, ethylacetate. Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 77301-42-9
Molecular Formula C13H14O2
Molecular Weight (g/mol) 202.25
MDL Number MFCD01632581
InChI Key OUVJWFRUESFCCY-SECBINFHSA-N
Synonym 1-6-methoxy-2-naphthyl ethanol, 1-6-methoxynaphthalen-2-yl ethanol, 1-6'-methoxy-2-naphthyl ethanol, +/--1-6-methoxynaphthyl ethanol, 2-1-hydroxyethyl-6-methoxynaphthalene, 1-6-methoxynaphthalen-2-yl ethan-1-ol, 6-methoxy-alpha-methyl-2-naphthalenemethanol, 1rs-1-6-methoxy-2-naphthyl ethanol naproxen impurity k, 1-6-methoxy-2-naphthyl ethan-1-ol, naproxen impurity k
PubChem CID 575523
SMILES COC1=CC=C2C=C(C=CC2=C1)[C@@H](C)O

Specifications

Melting Point 111°C to 112°C
Quantity 5 g
Solubility Information Soluble in DMSO,acetone,dichloromethane,ethylacetate. Insoluble in water.
IUPAC Name (1R)-1-(6-methoxynaphthalen-2-yl)ethan-1-ol
Formula Weight 202.25
Percent Purity 98%
Chemical Name or Material 1-(6-Methoxy-2-naphthyl)ethanol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.