missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediyl Bismethanethiosulfonate, TRC

Product Code. 30390751
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390751 10 mg -
30404209 50 mg 50mg
30394393 25 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390751 Supplier Toronto Research Chemicals Supplier No. TRCH29425010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-01-9
Molecular Formula C8 H18 O4 S4
Molecular Weight (g/mol) 306.49
IUPAC Name 1,6-bis(methylsulfonylsulfanyl)hexane
SMILES CS(=O)(=O)SCCCCCCSS(=O)(=O)C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C8H18O4S4/c1-15(9,10)13-7-5-3-4-6-8-14-16(2,11)12/h3-8H2,1-2H3
Formula Weight 306.0088
Chemical Name or Material 1,6-Hexanediyl Bismethanethiosulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.