missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol Dimethacrylate - stabilized with MEHQ, TRC

Product Code. 30396650
Change view
Click to view available options
Quantity:
500 mg
1 g
10 g
Unit Size:
10g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30396650 1 g -
30400554 500 mg -
30389437 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30396650 Supplier Toronto Research Chemicals Supplier No. TRCH2942481G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6606-59-3
Molecular Formula C14H22O4
Molecular Weight (g/mol) 254.32
IUPAC Name 6-(2-methylprop-2-enoyloxy)hexyl 2-methylprop-2-enoate
SMILES CC(=C)C(=O)OCCCCCCOC(=O)C(=C)C

Specifications

Quantity 1 g
InChI Formula InChI=1S/C14H22O4/c1-11(2)13(15)17-9-7-5-6-8-10-18-14(16)12(3)4/h1,3,5-10H2,2,4H3
Formula Weight 254.1518
Chemical Name or Material 1,6-Hexanediol Dimethacrylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.