missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], TRC

Product Code. 30397216
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30397216 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30397216 Supplier Toronto Research Chemicals Supplier No. TRCH296885100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Irganox 259
CAS 35074-77-2
Molecular Formula C40 H62 O6
Quantity 100 mg
Synonym Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 1,1'-(1,6-hexanediyl) ester, Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 1,6-hexanediyl ester (9CI), 1,6-Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxy]hexane, 1,6-Bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxy]hexane, 1,6-Hexamethylene bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), 1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 1,6-Hexanediol bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 1,6-Hexanediyl-bis(3,5-bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoate), Antioxidant 259, Hexamethylene 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Hexamethylene bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Hexamethylene bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Hexamethylene bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Hexamethylene glycol bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Hexamethylene glycol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Irganox 249, Irganox 259
InChI Formula InChI=1S/C40H62O6/c1-37(2,3)29-23-27(24-30(35(29)43)38(4,5)6)17-19-33(41)45-21-15-13-14-16-22-46-34(42)20-18-28-25-31(39(7,8)9)36(44)32(26-28)40(10,11)12/h23-26,43-44H,13-22H2,1-12H3
SMILES CC(C)(C)c1cc(CCC(=O)OCCCCCCOC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C
IUPAC Name 6-[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyloxy]hexyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate
Molecular Weight (g/mol) 638.92
Formula Weight 638.4546
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.