missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], TRC
Specifications
Specifications
| Chemical Name or Material | Irganox 259 |
| CAS | 35074-77-2 |
| Molecular Formula | C40 H62 O6 |
| Quantity | 10 mg |
| Synonym | Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 1,1'-(1,6-hexanediyl) ester, Benzenepropanoic acid, 3,5-bis(1,1-dimethylethyl)-4-hydroxy-, 1,6-hexanediyl ester (9CI), 1,6-Bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxy]hexane, 1,6-Bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionyloxy]hexane, 1,6-Hexamethylene bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), 1,6-Hexanediol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 1,6-Hexanediol bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], 1,6-Hexanediyl-bis(3,5-bis(1,1-dimethylethyl)-4-hydroxybenzenepropanoate), Antioxidant 259, Hexamethylene 3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate, Hexamethylene bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Hexamethylene bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Hexamethylene bis[β-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Hexamethylene glycol bis(3,5-di-tert-butyl-4-hydroxyhydrocinnamate), Hexamethylene glycol bis[3-(3,5-di-tert-butyl-4-hydroxyphenyl)propionate], Irganox 249, Irganox 259 |
| InChI Formula | InChI=1S/C40H62O6/c1-37(2,3)29-23-27(24-30(35(29)43)38(4,5)6)17-19-33(41)45-21-15-13-14-16-22-46-34(42)20-18-28-25-31(39(7,8)9)36(44)32(26-28)40(10,11)12/h23-26,43-44H,13-22H2,1-12H3 |
| SMILES | CC(C)(C)c1cc(CCC(=O)OCCCCCCOC(=O)CCc2cc(c(O)c(c2)C(C)(C)C)C(C)(C)C)cc(c1O)C(C)(C)C |
| IUPAC Name | 6-[3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoyloxy]hexyl 3-(3,5-ditert-butyl-4-hydroxyphenyl)propanoate |
| Molecular Weight (g/mol) | 638.92 |
| Formula Weight | 638.4546 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?