missing translation for 'onlineSavingsMsg'
Learn More

1,6-Hexanediol, 97%

Product Code. 11489653
Change view
Click to view available options
Quantity:
250 g
500 g
2500 g
Unit Size:
2500g
250g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11489653 500 g 500g
11464210 250 g 250g
11499653 2500 g 2500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11489653 Supplier Thermo Scientific Alfa Aesar Supplier No. A12439.36
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-11-8 | C6H14O2 | 118.176 g/mol

1,6-Hexanediol is used in polymer synthesis such as polyester, polyurethane and nylon. It is used as an intermediate to adhesives, acrylics and dyestuffs. Further, it is employed in gasoline refining and pharmaceutical production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Hexanediol is used in polymer synthesis such as polyester, polyurethane and nylon. It is used as an intermediate to adhesives, acrylics and dyestuffs. Further, it is employed in gasoline refining and pharmaceutical production.

Solubility
Soluble in water.

Notes
Hygroscopic. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, chloroformates and reducing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-11-8
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00002985
InChI Key XXMIOPMDWAUFGU-UHFFFAOYSA-N
Synonym 1,6-hexanediol, hexamethylene glycol, 1,6-dihydroxyhexane, hexamethylenediol, alpha,omega-hexanediol, .alpha.,.omega.-hexanediol, unii-zia319275i, ccris 8982, 6-hydroxy-1-hexanol, 1,6-hexylene glycol
PubChem CID 12374
ChEBI CHEBI:43078
IUPAC Name hexane-1,6-diol
SMILES C(CCCO)CCO

Specifications

Melting Point 40°C to 43°C
Density 1.120
Boiling Point 248°C to 250°C
Flash Point 147°C (296°F)
Assay Percent Range 97%
Quantity 500 g
Beilstein 1633461
Sensitivity Hygroscopic
Merck Index 14,4690
Solubility Information Soluble in water.
Formula Weight 118.18
Percent Purity 97%
Chemical Name or Material 1, 6-Hexanediol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.