missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ 1,6-Hexanediol
Description
1,6-Hexanediol is a widely used tool to probe liquid-liquid phase separation (LLPS) in cells. Disrupts hydrophobic interaction-induced phase separation assemblies both in vitro and in vivo. Dissolves LLPS condensates, such as P bodies, but not solid-like condensates, such as protein aggregates and cytoskeletal assemblies.Typically used as a 3-10% solution (w/v). Disrupts cellular structures in yeast and alters cellular morphology in HeLa cells.
Specifications
Specifications
| CAS | 629-11-8 |
| Quantity | 10 g |
| Product Line | Tocris |
| Molecular Formula | C6H14O2 |
| Purity | 98% |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?