missing translation for 'onlineSavingsMsg'
Learn More

1,6-Heptadiyne, 98%

Product Code. p-140074 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10383003

Brand: Thermo Scientific Acros 309420010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2396-63-6 | C7H8 | 92.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2396-63-6
Molecular Formula C7H8
Molecular Weight (g/mol) 92.15
MDL Number MFCD00014925
InChI Key RSPZSDWVQWRAEF-UHFFFAOYSA-N
Synonym 1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, acmc-1cin6, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
PubChem CID 337121
IUPAC Name hepta-1,6-diyne
SMILES C#CCCCC#C

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point -85°C
Color Yellow
Density 0.8050g/mL
Boiling Point 111.5°C
Flash Point 9°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula CH∽C(CH2)3C∽CH
Refractive Index 1.4400 to 1.444
Quantity 1 g
Specific Gravity 0.805
Solubility Information Solubility in water: insoluble
Formula Weight 92.15
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1, 6-Heptadiyne, 98%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.