missing translation for 'onlineSavingsMsg'
Learn More

1,6-Heptadiyne, 97%

Product Code. 11423714
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11423714 5 g 5g
11433714 25 g 25g
11443714 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11423714 Supplier Thermo Scientific Alfa Aesar Supplier No. A11318.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2396-63-6 | C7H8 | 92.141 g/mol

1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne). Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene. Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne). Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene. Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Haevy metal powder, heat. Refrigerate at (+4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2396-63-6
Molecular Formula C7H8
Molecular Weight (g/mol) 92.141
MDL Number MFCD00014925
InChI Key RSPZSDWVQWRAEF-UHFFFAOYSA-N
Synonym 1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, acmc-1cin6, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
PubChem CID 337121
IUPAC Name hepta-1,6-diyne
SMILES C#CCCCC#C

Specifications

Density 0.81
Boiling Point 109°C to 111°C
Flash Point 9°C (48°F)
Odor Recognizable
Refractive Index 1.442
Quantity 5 g
UN Number UN3295
Beilstein 1731756
Solubility Information Insoluble in water.
Formula Weight 92.14
Percent Purity 97%
Chemical Name or Material 1,6-Heptadiyne
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.