missing translation for 'onlineSavingsMsg'
Learn More

1,6-Heptadiyne, 97%

Product Code. 11433714
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11433714

Brand: Thermo Scientific Alfa Aesar A11318.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2396-63-6 | C7H8 | 92.141 g/mol

1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne). Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene. Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Heptadiyne may be used in the preparation of free-standing polymer films of poly(1,6-heptadiyne). Polymerization of 1,6-heptadiyne using an insoluble Zeigler-Natta catalyst affords soluble polymer having six-membered ring containing polyene. Ruthenium(II)-catalyzed reaction of a substituted 1,6-heptadiyne with norbornene affords a tandem [2+ 2+ 2]/[4+2] cycloaddition product and a [2+ 2+2] cycloadduct.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Haevy metal powder, heat. Refrigerate at (+4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2396-63-6
Molecular Formula C7H8
Molecular Weight (g/mol) 92.141
MDL Number MFCD00014925
InChI Key RSPZSDWVQWRAEF-UHFFFAOYSA-N
Synonym 1,6-heptadiyne, 1,6-heptadiyne 6ci, 8ci, 9ci, heptane-1,6-diyne, acmc-1cin6, 1,6-heptadiyne, 97+%, wln: 1uu5uu1, 4-01-00-01121 beilstein handbook reference
PubChem CID 337121
IUPAC Name hepta-1,6-diyne
SMILES C#CCCCC#C

Specifications

Density 0.81
Boiling Point 109°C to 111°C
Flash Point 9°C (48°F)
Odor Recognizable
Refractive Index 1.442
Quantity 25 g
UN Number UN3295
Beilstein 1731756
Solubility Information Insoluble in water.
Formula Weight 92.14
Percent Purity 97%
Chemical Name or Material 1,6-Heptadiyne
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.