missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Ethoxy-2-pyridyl)piperazine, 97%, Thermo Scientific Chemicals

Catalog Number 11479610
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
108122-25-4
C11H17N3O
207.277
MFCD08061051
IGBPLIGXKQSNKV-UHFFFAOYSA-N
13795742
1-(6-ethoxypyridin-2-yl)piperazine
CCOC1=CC=CC(=N1)N2CCNCC2
This item is not returnable. View return policy
108122-25-4
C11H17N3O
207.277
MFCD08061051
IGBPLIGXKQSNKV-UHFFFAOYSA-N
13795742
1-(6-ethoxypyridin-2-yl)piperazine
CCOC1=CC=CC(=N1)N2CCNCC2

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Quantity 250 mg
Synonym 1-6-ethoxypyridin-2-yl piperazine, 1-6-ethoxy-2-pyridyl piperazine, piperazine, 1-6-ethoxy-2-pyridinyl, acmc-20ap9w, 2-ethoxy-6-piperazinylpyridine, 1-6-ethoxy-pyridin-2-yl-piperazine, piperazine,1-6-ethoxy-2-pyridinyl
Formula Weight 207.28
Percent Purity 97%
Chemical Name or Material 1-(6-Ethoxy-2-pyridyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'