missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 1,6-Diisocyanatohexane, 98+%

Brand:  Alfa Aesar™ 043803.30

Code : NEW

Additional Details : CAS Number : 822-06-0 Weight : 0.25000kg Transport : UN number : 2281 Chem class : 6.1 Pack group : II

Quantity 250g

 View more versions of this product

Product Code. 11330448

  • £77.90 / 250g


EU Stock 5
Add to basket

Description

Description

  • Packaged under Argon
Specifications

Chemical Identifiers

1,6-Diisocyanatohexane
OCN(CH2)6NCO
MFCD00002047
956709
822-06-0
168.19
UN2281
Hexamethylene diisocyanate

Specifications

1,6-Diisocyanatohexane
1.05g/mL
−67°C
≥98%
OCN(CH2)6NCO
130°C (266°F)
168.19
Moisture Sensitive
956709
822-06-0
1.4520
Liquid
255°C
250g
Pungent
MFCD00002047
UN2281
Hexamethylene diisocyanate
Videos
SDS