missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dicyanohexane, 99%

Product Code. 10010870 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mL
Packaging:
Glass bottle
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10010870 100 mL Glass bottle 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10010870 Supplier Thermo Scientific Acros Supplier No. 113861000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-40-3 | C8H12N2 | 136.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 629-40-3
CAS Min % 98.5
CAS Max % 100.0
Melting Point -3.5°C
Color Yellow
Density 0.9500g/mL
Boiling Point 175.0°C (10.0 mmHg)
Flash Point 175°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Molecular Formula C8H12N2
Linear Formula NC(CH2)6CN
Refractive Index 1.4430 to 1.4450
MDL Number MFCD00001981
Quantity 100 mL
Beilstein 02, 694
Specific Gravity 0.95
Synonym 1,6-dicyanohexane, suberonitrile, suberic acid dinitrile, suberodinitrile, unii-jnt1lxa02u, korksaeuredinitril german, jnt1lxa02u, hexamethylene di-isocyanide, hexane-1,6-dicarbonitrile, hexamethylenediisocyanic acid
Solubility Information Solubility in water: 1.2%
InChI Key BTNXBLUGMAMSSH-UHFFFAOYSA-N
SMILES C(CCCC#N)CCC#N
IUPAC Name octanedinitrile
Molecular Weight (g/mol) 136.2
PubChem CID 12385
Viscosity 8.5 mPa.s (20°C)
Formula Weight 136.2
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1, 6-Dicyanohexane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,6-Dicyanohexane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
Hazard Statement
  • H301-Toxic if swallowed.
Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.