missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dichlorohexane, 98%

Product Code. 11430387
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11430387 500 g 500g
11420387 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11430387 Supplier Thermo Scientific Alfa Aesar Supplier No. B21763.36
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2163-00-0 | C6H12Cl2 | 155.062 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2163-00-0
Molecular Formula C6H12Cl2
Molecular Weight (g/mol) 155.062
MDL Number MFCD00001020
InChI Key OVISMSJCKCDOPU-UHFFFAOYSA-N
Synonym hexane, 1,6-dichloro, hexamethylene dichloride, hexamethylene chloride, 1,6-dichlorhexan, hexane,6-dichloro, 1,6-dichloro-hexan, 1,6-dichloro hexane, hexane,1,6-dichloro, acmc-209fml
PubChem CID 16551
IUPAC Name 1,6-dichlorohexane
SMILES C(CCCCl)CCCl

Specifications

Melting Point -13°C
Density 1.067
Boiling Point 204°C to 205°C
Flash Point 73°C (163°F)
Refractive Index 1.4568
Quantity 500 g
UN Number UN3082
Beilstein 773673
Formula Weight 155.07
Percent Purity 98%
Chemical Name or Material 1,6-Dichlorohexane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.