missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dichlorohexane, 98%, ACROS Organics™

Product Code. 10684952 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
100g; Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
10684952 100g; Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10684952 Supplier Thermo Scientific Acros Supplier No. 113411000

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 2163-00-0
CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.0700g/mL
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Molecular Formula C6H12Cl2
Linear Formula Cl(CH2)6Cl
Refractive Index 1.4558 to 1.4578 (20°C, 589nm)
MDL Number MFCD00001020
Quantity 100 g
Beilstein 01, 144
Synonym hexane, 1,6-dichloro, hexamethylene dichloride, hexamethylene chloride, 1,6-dichlorhexan, hexane,6-dichloro, 1,6-dichloro-hexan, 1,6-dichloro hexane, hexane,1,6-dichloro, acmc-209fml
InChI Key OVISMSJCKCDOPU-UHFFFAOYSA-N
SMILES C(CCCCl)CCCl
IUPAC Name 1,6-dichlorohexane
Molecular Weight (g/mol) 155.062
PubChem CID 16551
Formula Weight 155.07
Percent Purity ≥97.5%
Physical Form Liquid
Chemical Name or Material 1, 6-Dichlorohexane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.