missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,6-Dibromohexane, TRC
Click to view available options
Quantity:
1 Kg
Unit Size:
1kg
Specifications
Specifications
| Chemical Name or Material | 1,6-Dibromohexane |
| CAS | 629-03-8 |
| Molecular Formula | C6 H12 Br2 |
| Quantity | 1 Kg |
| Synonym | Hexane, 1,6-dibromo-, 1,6-Dibromohexane, 1,6-Dibromo-n-hexane, Hexamethylene dibromide, NSC 7306, α,ω-Dibromohexane |
| InChI Formula | InChI=1S/C6H12Br2/c7-5-3-1-2-4-6-8/h1-6H2 |
| SMILES | BrCCCCCCBr |
| IUPAC Name | 1,6-dibromohexane |
| Molecular Weight (g/mol) | 243.967 |
| Formula Weight | 241.9306 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction