missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromohexane, 97+%

Product Code. p-7021185
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11485443

Brand: Thermo Scientific Alfa Aesar A13417.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 629-03-8 | C6H12Br2 | 243.97 g/mol

1,6-Dibromohexane is used as a reagent in the synthesis of novel benzo[b]xanthone derivatives which have a potential antitumor activity. It is also used as a cross-linker for the cross-linking of glycuronans.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,6-Dibromohexane is used as a reagent in the synthesis of novel benzo[b]xanthone derivatives which have a potential antitumor activity. It is also used as a cross-linker for the cross-linking of glycuronans.

Solubility
Miscible with ethanol, ether, benzene and chloroform. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-03-8
Molecular Formula C6H12Br2
Molecular Weight (g/mol) 243.97
MDL Number MFCD00000272
InChI Key SGRHVVLXEBNBDV-UHFFFAOYSA-N
Synonym hexamethylene dibromide, hexane, 1,6-dibromo, 1,6-dibromo-n-hexane, 1,6-dibromohexan, alpha,omega-dibromohexane, 1,6-dibromohexan german, hexamethylene bromide, labotest-bb ltbb001557, 1,6 dibromohexane
PubChem CID 12368
IUPAC Name 1,6-dibromohexane
SMILES C(CCCBr)CCBr

Specifications

Melting Point -2°C
Density 1.61
Boiling Point 242°C to 244°C
Flash Point >110°C (230°F)
Odor Odorless
Refractive Index 1.5075
Quantity 100 g
UN Number UN3082
Beilstein 1236322
Solubility Information Miscible with ethanol,ether,benzene and chloroform. Immiscible with water.
Formula Weight 243.98
Percent Purity ≥97%
Chemical Name or Material 1,6-Dibromohexane
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.