missing translation for 'onlineSavingsMsg'
Learn More

1,6-Dibromo-2-naphthol, 99%, Thermo Scientific™

Product Code. 10182230 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10182230 100 g 100g
10635532 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10182230 Supplier Thermo Scientific Acros Supplier No. 112851000
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 16239-18-2
Molecular Formula C10H6Br2O
Molecular Weight (g/mol) 301.97
MDL Number MFCD00003870
InChI Key VKESFYLPKHQOOA-UHFFFAOYSA-N
Synonym 1,6-dibromo-2-naphthol, 2-naphthalenol, 1,6-dibromo, 2-naphthol, 1,6-dibromo, 1,6-dibromo-2-hydroxynaphthalene, 1,6-dibromo-beta-naphthol, 1,6-dibromo-.beta.-naphthol, pubchem22920, 2-naphthol,6-dibromo, 1,6 dibromo-2-napthol, acmc-1c8jh
PubChem CID 27768
IUPAC Name 1,6-dibromonaphthalen-2-ol
SMILES OC1=CC=C2C=C(Br)C=CC2=C1Br

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 105.0°C to 107.0°C
Color Brown or Pink to Purple
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula Br2C10H5OH
Quantity 100 g
Beilstein 06, 652
Formula Weight 301.96
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 1, 6-Dibromo-2-naphthol, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 1,6-Dibromo-2-naphthol
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.