missing translation for 'onlineSavingsMsg'
Learn More

1,6-Diaminohexane, TRC

Product Code. 30041312
Change view
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30041312 50 g 50g
30041290 100 g 100g
30007482 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30041312 Supplier Toronto Research Chemicals Supplier No. TRCD41618050G
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 124-09-4
Molecular Formula C6 H16 N2
Molecular Weight (g/mol) 116.2046
Synonym 1,6-Hexanediamine, 1,6-Diamino-n-hexane, 1,6-Diaminohexane, 1,6-Hexylenediamine, 6-Aminohexylamine, 80HMD, Advancure, Dytek 70, Dytek HMD, HMDA, Hexamethylenediamine, Hexylenediamine, Hi Perm, NSC 9257, RT Advancure HD, V 1, α,ω-Hexanediamine, Hexane-1,6-diamine
IUPAC Name hexane-1,6-diamine
SMILES NCCCCCCN

Specifications

Quantity 50 g
InChI Formula InChI=1S/C6H16N2/c7-5-3-1-2-4-6-8/h1-8H2
Formula Weight 116.1313
Chemical Name or Material Hexane-1,6-diamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.