missing translation for 'onlineSavingsMsg'
Learn More

1-(6-Chloro-2-oxo-4-(trifluoromethyl)-2,4-dihydro-1H-benzo[d][1,3]oxazin-4-yl)pent-1-yn-3-yl Methanesulfonate, TRC

Product Code. 30357726
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30357726 1 mg 1mg
30359863 5 mg 5mg
30366665 2.5 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30357726 Supplier Toronto Research Chemicals Supplier No. TRCC2145151MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C15 H13 Cl F3 N O5 S
Molecular Weight (g/mol) 411.781
IUPAC Name 1-[6-chloro-2-oxo-4-(trifluoromethyl)-1H-3,1-benzoxazin-4-yl]pent-1-yn-3-yl methanesulfonate
SMILES CCC(OS(=O)(=O)C)C#CC1(OC(=O)Nc2ccc(Cl)cc12)C(F)(F)F

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C15H13ClF3NO5S/c1-3-10(25-26(2,22)23)6-7-14(15(17,18)19)11-8-9(16)4-5-12(11)20-13(21)24-14/h4-5,8,10H,3H2,1-2H3,(H,20,21)
Formula Weight 411.016
Chemical Name or Material 1-(6-Chloro-2-oxo-4-(trifluoromethyl)-2,4-dihydro-1H-benzo[d][1,3]oxazin-4-yl)pent-1-yn-3-yl Methanesulfonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.