missing translation for 'onlineSavingsMsg'
Learn More

1,6-Bismaleimidohexane, 97%

Product Code. 15465058
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 15465058

Brand: Thermo Scientific Alfa Aesar H58874.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 4856-87-5 | C14H16N2O4 | 276.292 g/mol

A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
A sulfhydryl reactive homobifunctional cross linking reagent. Under mild conditions, will permit irreversible cross-linking of sulfhydral containing compounds at a pH range of 6.5 to 7.5.

Solubility
Soluble in chloroform.

Notes
Protect from heat. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4856-87-5
Molecular Formula C14H16N2O4
Molecular Weight (g/mol) 276.292
MDL Number MFCD00047122
InChI Key PYVHLZLQVWXBDZ-UHFFFAOYSA-N
Synonym 1,6-bismaleimidohexane, 1,6-dimaleimidohexane, n,n'-hexamethylenedimaleimide, 1,6-bis maleimido hexane, 1,1'-hexane-1,6-diyl bis 1h-pyrrole-2,5-dione, 1h-pyrrole-2,5-dione, 1,1'-1,6-hexanediyl bis, maleimide, n,n'-hexamethylenedi, n,n'-hexamethylenebis maleimide, 1,1'-1,6-hexanediyl bis-1h-pyrrole-2,5-dione
PubChem CID 20992
IUPAC Name 1-[6-(2,5-dioxopyrrol-1-yl)hexyl]pyrrole-2,5-dione
SMILES C1=CC(=O)N(C1=O)CCCCCCN2C(=O)C=CC2=O

Specifications

Melting Point 144°C
Quantity 100 mg
Solubility Information Soluble in chloroform.
Formula Weight 276.29
Percent Purity 97%
Chemical Name or Material 1,6-Bismaleimidohexane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.