missing translation for 'onlineSavingsMsg'
Learn More

1,6,7-Trimethylnaphthalene, TRC

Product Code. 30457141
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30457141 10 mg
30451690 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30457141 Supplier Toronto Research Chemicals Supplier No. TRCT79647510MG
 Restricted Item
Estimated Shipment: 10-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2245-38-7
Molecular Formula C13 H14
Molecular Weight (g/mol) 170.25
Synonym Naphthalene, 1,6,7-trimethyl- (6CI, 7CI, 8CI, 9CI, ACI), 1,6,7-Trimethylnaphthalene (ACI), 2,3,5-Trimethylnaphthalene, NSC 89511
IUPAC Name 1,6,7-trimethylnaphthalene
SMILES Cc1cc2cccc(C)c2cc1C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C13H14/c1-9-5-4-6-12-7-10(2)11(3)8-13(9)12/h4-8H,1-3H3
Formula Weight 170.1096
Chemical Name or Material 2,3,5-Trimethylnaphthalene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.