missing translation for 'onlineSavingsMsg'
Learn More

1-(5-(Trifluoromethyl)-2-pyridyl)piperidine-4-carboxylic acid, 95%, Thermo Scientific™

Product Code. 10317164
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10317164

Brand: Thermo Scientific Maybridge KM07694EA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 406476-31-1
Molecular Formula C12H13F3N2O2
Molecular Weight (g/mol) 274.24
MDL Number MFCD00209248
InChI Key KNDSIDUPVUCATQ-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylic acid, 1-5-trifluoromethyl-2-pyridyl piperidine-4-carboxylic acid, 5'-trifluoromethyl-3,4,5,6-tetrahydro-2h-1,2' bipyridinyl-4-carboxylic acid, peakdale1_002677, 2-4-carboxypiperidin-1-yl-5-trifluoromethyl pyridine, 4-carboxy-1-5-trifluoromethyl pyridin-2-yl piperidine, 1-5-trifluoromethyl pyridin-2-yl piperidine-4-carboxylicacid, 1-5-trifluoromethylpyrid-2-yl-piperidine-4-carboxylic acid
PubChem CID 2777788
SMILES [O-]C(=O)C1CCN(CC1)C1=CC=C(C=[NH+]1)C(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%
Quantity 10 g
IUPAC Name 1-[5-(trifluoromethyl)pyridin-2-yl]piperidine-4-carboxylic acid
Formula Weight 274.24
Percent Purity 95%
Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridyl]piperidine-4-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.