missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(5-(Trifluoromethyl)-2-pyridyl)piperidine-4-carboxylic acid, 95%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
10 g
Specifications
CAS Min % | 95.0 |
Assay Percent Range | 95% |
Packaging | Amber glass bottle |
Quantity | 250 mg |
IUPAC Name | 2-(4-carboxylatopiperidin-1-yl)-5-(trifluoromethyl)pyridin-1-ium |
Formula Weight | 274.24 |
Percent Purity | 95% |
Chemical Name or Material | 1-[5-(Trifluoromethyl)-2-pyridyl]piperidine-4-carboxylic acid |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'