missing translation for 'onlineSavingsMsg'
Learn More

1-[5-(Trifluoromethyl)-2-pyridyl]-1,4-diazepane,95+%, Thermo Scientific™

Product Code. 10308483
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10308483 1 g Amber Glass Bottle 1g
10680656 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10308483 Supplier Thermo Scientific Maybridge Supplier No. AW00291DA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 306934-70-3
Molecular Formula C11H14F3N3
Molecular Weight (g/mol) 245.25
MDL Number MFCD00661783
InChI Key IBMSHQVIAUTKDL-UHFFFAOYSA-N
Synonym 1-5-trifluoromethyl pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazepane, 1-5-trifluoromethyl pyrid-2-yl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4 diazepane, 1h-1,4-diazepine,hexahydro-1-5-trifluoromethyl-2-pyridinyl, 1-5-trifluoromethyl-pyridin-2-yl 1,4-diazepane, 1-5-trifluoromethyl-2-pyridinyl-1,4-diazepane, 1-5-trifluoromethyl-pyridin-2-yl-1,4-diazepane, 1-5-trifluoromethyl-2-pyridyl-1,4-diazaperhydroepine
PubChem CID 2760779
IUPAC Name 1-[5-(trifluoromethyl)pyridin-2-yl]-1,4-diazepane
SMILES FC(F)(F)C1=CN=C(C=C1)N1CCCNCC1

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 245.25
Percent Purity 97%
Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridyl]-1, 4-diazepane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.