missing translation for 'onlineSavingsMsg'
Learn More

1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine, 98%, Thermo Scientific™

Product Code. 10088911 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10088911 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 10088911 Supplier Thermo Scientific Acros Supplier No. 367100010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 132834-58-3
Molecular Formula C10H14F3N3
Molecular Weight (g/mol) 233.24
MDL Number MFCD00114708
InChI Key BNMSJUIMZULLAS-UHFFFAOYSA-P
Synonym 1-5-trifluoromethyl pyridin-2-yl piperazine, 1-5-trifluoromethyl pyrid-2-yl piperazine, 1-5-trifluoromethyl-pyridin-2-yl-piperazine, 1-5-trifluoromethyl-2-pyridinyl piperazine, 1-5-trifluoromethyl-2-pyridinyl-piperazine, 1-5-trifluoromethyl-2-pyridyl piperazine, 1-5-trifluoromethyl pyridine-2-yl piperazine, piperazine, 1-5-trifluoromethyl-2-pyridinyl, 5-trifluoromethyl-2-pyridyl piperazine
PubChem CID 667618
SMILES FC(F)(F)C1=CC=C([NH+]=C1)N1CC[NH2+]CC1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Yellow
Boiling Point 122.0°C (2.0 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 1 g
IUPAC Name 1-[5-(trifluoromethyl)pyridin-2-yl]piperazine
Formula Weight 231.22
Percent Purity 98%
Physical Form Crystalline Chunks
Chemical Name or Material 1-[5-(Trifluoromethyl)-2-pyridinyl]piperazine, 98%
Show More Show Less
compliance-icons
Product Identifier
  • 1-¢5-(Trifluoromethyl)-2-pyridinyl!piperazine
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.