missing translation for 'onlineSavingsMsg'
Learn More

1,5-Pentanedithiol, 96%

Product Code. 11411687
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11411687 5 g 5g
11421687 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11411687 Supplier Thermo Scientific Alfa Aesar Supplier No. B25665.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 928-98-3 | C5H12S2 | 136.27 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-98-3
Molecular Formula C5H12S2
Molecular Weight (g/mol) 136.27
MDL Number MFCD00004908
InChI Key KMTUBAIXCBHPIZ-UHFFFAOYSA-N
Synonym 1,5-pentanedithiol, 1,5-dimercaptopentane, unii-r8rc3q7t1w, pentamethylene dimercaptan, r8rc3q7t1w, pentamethylenedithiol, 1,5-pentanodithiol, acmc-209rih, pentamethylenedimercaptane, 1,5-pentanedithiol 98+%
PubChem CID 70236
IUPAC Name pentane-1,5-dithiol
SMILES SCCCCCS

Specifications

Melting Point -72°C
Boiling Point 216°C to 218°C
Flash Point 95°C (203°F)
Odor Unpleasant
Quantity 5 g
UN Number UN3334
Beilstein 1732335
Formula Weight 136.28
Percent Purity 96%
Chemical Name or Material 1,5-Pentanedithiol

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.