missing translation for 'onlineSavingsMsg'
Learn More

1,5-Naphthyridine, 97+%

Product Code. 15491709
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15491709 5 g 5g
15401719 250 mg 250mg
15481709 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15491709 Supplier Thermo Scientific Alfa Aesar Supplier No. H64484.06
 Restricted Item
Estimated Shipment: 23-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 254-79-5 | C8H6N2 | 130.15 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 254-79-5
Molecular Formula C8H6N2
Molecular Weight (g/mol) 130.15
MDL Number MFCD00039727
InChI Key VMLKTERJLVWEJJ-UHFFFAOYSA-N
Synonym 1,5-diazanaphthalene, 1,5-pyridopyridine, pyridino 3,2-b pyridine, pyridopyridine, 1,5-naphtyridine, 1,5 naphthyridine, zlchem 698, pubchem18072, acmc-209gka, pyrido 1,5-b pyridine
PubChem CID 136070
ChEBI CHEBI:36625
IUPAC Name 1,5-naphthyridine
SMILES C1=CC2=NC=CC=C2N=C1

Specifications

Melting Point 69°C to 75°C
Color White
Density 1.25 g/mL
Quantity 5 g
Beilstein 109350
Formula Weight 130.15
Percent Purity ≥97%
Physical Form Solid
Chemical Name or Material 1,5-Naphthyridine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.