missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Isoquinolinesulfonyl)piperazine Hydrochloride, TRC

Product Code. 30400948
Change view
Click to view available options
Quantity:
25 mg
50 mg
250 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30400948 50 mg
30406893 25 mg
30403759 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30400948 Supplier Toronto Research Chemicals Supplier No. TRCI89100050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 141543-63-7
Molecular Formula C13 H15 N3 O2 S . Cl H
Molecular Weight (g/mol) 313.8
IUPAC Name 5-piperazin-1-ylsulfonylisoquinoline;hydrochloride
SMILES Cl.O=S(=O)(N1CCNCC1)c2cccc3cnccc23

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C13H15N3O2S.ClH/c17-19(18,16-8-6-14-7-9-16)13-3-1-2-11-10-15-5-4-12(11)13;/h1-5,10,14H,6-9H2;1H
Formula Weight 313.0652
Chemical Name or Material 1-(5-Isoquinolinesulfonyl)piperazine, Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.