missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Isoquinolinesulfonyl)-2-methylpiperazine Dihydrochloride, TRC

Product Code. 30402602
Change view
Click to view available options
Quantity:
5 mg
250 mg
Unit Size:
5mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30402602 5 mg 5mg
30390049 250 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30402602 Supplier Toronto Research Chemicals Supplier No. TRCI8850005MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 108930-17-2
Molecular Formula C14 H17 N3 O2 S . 2 Cl H
Molecular Weight (g/mol) 364.29
IUPAC Name 5-(2-methylpiperazin-1-yl)sulfonylisoquinoline;dihydrochloride
SMILES Cl.Cl.CC1CNCCN1S(=O)(=O)c2cccc3cnccc23

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C14H17N3O2S.2ClH/c1-11-9-16-7-8-17(11)20(18,19)14-4-2-3-12-10-15-6-5-13(12)14;;/h2-6,10-11,16H,7-9H2,1H3;2*1H
Formula Weight 363.0575
Chemical Name or Material 1-(5-Isoquinolinesulfonyl)-2-methylpiperazine dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.