missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Iodonaphthalene-1-sulfonyl)-1H-hexahydro-1,4 -diazepine, Hydrochloride, TRC

Product Code. 30399269
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399269 50 mg -
30407680 10 mg -
30403651 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30399269 Supplier Toronto Research Chemicals Supplier No. TRCI71500050MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 110448-33-4
Molecular Formula C15 H17 I N2 O2 S . Cl H
Molecular Weight (g/mol) 452.74
Synonym ML 7 hydrochloride
IUPAC Name 1-(5-iodonaphthalen-1-yl)sulfonyl-1,4-diazepane;hydrochloride
SMILES Cl.Ic1cccc2c(cccc12)S(=O)(=O)N3CCCNCC3

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C15H17IN2O2S.ClH/c16-14-6-1-5-13-12(14)4-2-7-15(13)21(19,20)18-10-3-8-17-9-11-18;/h1-2,4-7,17H,3,8-11H2;1H
Formula Weight 451.9822
Chemical Name or Material 1-(5-Iodonaphthalene-1-sulfonyl)-1H-hexahydro-1,4 -diazepine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.