missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Hexadiene, 98%
Click to view available options
Quantity:
25 g
100 g
500 g
Packungsgröße:
100g
25g
500g
Beschreibung
1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.
Solubility
Soluble in water(0.17g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.
Solubility
Soluble in water(0.17g/L).
Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Spezifikation
Spezifikation
| Melting Point | -141°C |
| Density | 0.692 |
| Boiling Point | 58°C to 60°C |
| Flash Point | −27°C (−16°F) |
| Odor | Turpentine-like |
| Refractive Index | 1.404 |
| Quantity | 25 g |
| UN Number | UN2458 |
| Beilstein | 1731106 |
| Solubility Information | Soluble in water(0.17g/L). |
| Mehr anzeigen |
Sicherheit und Handhabung
Produktidentifikator
- 1,5-Hexadiene
- Danger
- Aspiration toxicity Category 1
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H304-May be fatal if swallowed and enters airways.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : 1,5-Hexadiene
RUO – Research Use Only
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur