missing translation for 'onlineSavingsMsg'
Learn More

1,5-Hexadiene, 98%

Product Code. 11470234
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11470234 25 g 25g
11480234 100 g 100g
11490234 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11470234 Supplier Thermo Scientific Alfa Aesar Supplier No. A13650.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 592-42-7 | C6H10 | 82.146 g/mol

1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Hexadiene is used to study the Pd(II)-catalyzed cycloisomerization of functionalized 1,5-hexadienes.

Solubility
Soluble in water(0.17g/L).

Notes
Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 592-42-7
Molecular Formula C6H10
Molecular Weight (g/mol) 82.146
MDL Number MFCD00008666
InChI Key PYGSKMBEVAICCR-UHFFFAOYSA-N
Synonym 1,5-hexadiene, biallyl, diallyl, hexadiene dot, alpha,omega-hexadiene, .alpha.,.omega.-hexadiene, unii-4mtz4764fi, 1,5-hexanediene, acmc-1axer, hexa-1,5-dien-1-yl
PubChem CID 11598
IUPAC Name hexa-1,5-diene
SMILES C=CCCC=C

Specifications

Melting Point -141°C
Density 0.692
Boiling Point 58°C to 60°C
Flash Point −27°C (−16°F)
Odor Turpentine-like
Refractive Index 1.404
Quantity 25 g
UN Number UN2458
Beilstein 1731106
Solubility Information Soluble in water(0.17g/L).
Formula Weight 82.15
Percent Purity 98%
Chemical Name or Material 1,5-Hexadiene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 1,5-Hexadiene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contain: 1,5-Hexadiene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.