missing translation for 'onlineSavingsMsg'
Learn More

1-(5-Fluoro-2-hydroxyphenyl)ethan-1-one, 97%, Thermo Scientific™

Product Code. 10031134
Change view
Click to view available options
Quantity:
10 g
50 g
100 g
Unit Size:
100g
10g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10031134 50 g 50g
10567644 10 g 10g
10294684 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10031134 Supplier Thermo Scientific Maybridge Supplier No. BTB04130EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 394-32-1
Molecular Formula C8H7FO2
Molecular Weight (g/mol) 154.14
InChI Key KOFFXZYMDLWRHX-UHFFFAOYSA-N
Synonym 5'-fluoro-2'-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl ethanone, 1-5-fluoro-2-hydroxyphenyl ethan-1-one, 5-fluoro-2-hydroxyacetophenone, 1-5-fluoro-2-hydroxyphenyl-1-ethanone, 2-hydroxy-5-fluoroacetophenone, 2'-hydroxy-5'-fluoroacetophenone, ethanone, 1-5-fluoro-2-hydroxyphenyl, 5'-fluoro-2'-hydroxy acetophenone, 5'-fluoro-2'-hydroxy-acetophenone
PubChem CID 95992
IUPAC Name 1-(5-fluoro-2-hydroxyphenyl)ethanone
SMILES CC(=O)C1=C(C=CC(=C1)F)O

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula FC6H3(OH)COCH3
Quantity 50 g
Formula Weight 154.14
Percent Purity 97%
Chemical Name or Material 1-(5-Fluoro-2-hydroxyphenyl)ethan-1-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.