missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Diphenylcarbohydrazide, Powder, BAKER ANALYZED™ A.C.S. Reagent, J.T. Baker™
High quality chemicals for laboratory and specialized industrial use
£256.00
Chemical Identifiers
CAS | 140-22-7 |
---|---|
Molecular Formula | C13H14N4O |
Molecular Weight (g/mol) | 242.282 |
InChI Key | KSPIHGBHKVISFI-UHFFFAOYSA-N |
Synonym | 1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl |
PubChem CID | 8789 |
ChEBI | CHEBI:4641 |
IUPAC Name | 1,3-dianilinourea |
SMILES | C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
Chemical Identifiers
140-22-7 | |
242.282 | |
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
CHEBI:4641 | |
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 |
C13H14N4O | |
KSPIHGBHKVISFI-UHFFFAOYSA-N | |
8789 | |
1,3-dianilinourea |
Specifications
140-22-7 | |
1,5-diphenylcarbazide, diphenylcarbazide, 1,5-diphenylcarbohydrazide, carbonic dihydrazide, 2,2'-diphenyl, 1,5-diphenylcarbonohydrazide, 1,5-diphenylcabohydrazide, sym-diphenylcarbazide, n,n'-diphenylcarbazide, 2,2'-diphenylcarbazide, carbohydrazide, 1,5-diphenyl | |
C1=CC=C(C=C1)NNC(=O)NNC2=CC=CC=C2 | |
242.282 | |
CHEBI:4641 | |
BAKER ANALYZED™ ACS Reagent | |
1,5-Diphenylcarbohydrazide |
C13H14N4O | |
KSPIHGBHKVISFI-UHFFFAOYSA-N | |
1,3-dianilinourea | |
8789 | |
242.28 | |
Powder |