missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diphenylcarbazone Compound With 1,5-Diphenylcarbazide (Technical Grade), TRC

Product Code. 30374653
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30374653 50 g
30376756 10 g
30382506 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30374653 Supplier Toronto Research Chemicals Supplier No. TRCD48735350G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 538-62-5
Molecular Formula C13 H12 N4 O
Molecular Weight (g/mol) 240.261
IUPAC Name (3E)-1-anilino-3-phenyliminourea
SMILES O=C(NNc1ccccc1)\N=N\c2ccccc2

Specifications

Quantity 50 g
InChI Formula InChI=1S/C13H12N4O/c18-13(16-14-11-7-3-1-4-8-11)17-15-12-9-5-2-6-10-12/h1-10,14H,(H,16,18)/b17-15+
Formula Weight 240.101
Chemical Name or Material 1,5-Diphenylcarbazone Compound With 1,5-Diphenylcarbazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.