missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethylhexylamine Hydrochloride, TRC

Product Code. 30387292
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30387292 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30387292 Supplier Toronto Research Chemicals Supplier No. TRCD4731965G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Octodrine hydrochloride
CAS 5984-59-8
Purity Grade Notes HPLC
Molecular Formula C8 H19 N . Cl H
Quantity 5 g
Synonym 2-Heptanamine, 6-methyl-, hydrochloride (1:1), 2-Heptanamine, 6-methyl-, hydrochloride (9CI), Hexylamine, 1,5-dimethyl-, hydrochloride (8CI), 2-Amino-6-methylheptane hydrochloride, 2-Isooctylamine hydrochloride, NSC 27116, 6-Methylheptan-2-amine hydrochloride, Octodrine hydrochloride, 6-Methyl-2-heptanamine hydrochloride
InChI Formula InChI=1S/C8H19N.ClH/c1-7(2)5-4-6-8(3)9;/h7-8H,4-6,9H2,1-3H3;1H
SMILES Cl.CC(C)CCCC(C)N
IUPAC Name 6-methylheptan-2-amine;hydrochloride
Molecular Weight (g/mol) 165.7
Formula Weight 165.1284
Percent Purity >95
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.