missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethyl Pentanedioate, TRC

Product Code. 30376531
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30376531 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30376531 Supplier Toronto Research Chemicals Supplier No. TRCD9991201G
 Restricted Item
Estimated Shipment: 20-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Glutaric acid dimethyl ester
CAS 1119-40-0
Molecular Formula C7 H12 O4
Quantity 1 g
Synonym Pentanedioic acid, 1,5-dimethyl ester, Glutaric acid, dimethyl ester (6CI,7CI,8CI), Pentanedioic acid, dimethyl ester (9CI), 1,5-Dimethyl pentanedioate, DBE 5, Dimethyl 1,3-propanedicarboxylate, Dimethyl glutarate, Dimethyl pentanedioate, Methyl glutarate, NSC 58578, Glutaric acid dimethyl ester
InChI Formula InChI=1S/C7H12O4/c1-10-6(8)4-3-5-7(9)11-2/h3-5H2,1-2H3
SMILES COC(=O)CCCC(=O)OC
IUPAC Name dimethyl pentanedioate
Molecular Weight (g/mol) 160.17
Formula Weight 160.0736
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.