missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethyl Citrate, TRC

Product Code. 30382911
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382911 25 g -
30384848 10 g -
30382345 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382911 Supplier Toronto Research Chemicals Supplier No. TRCD47086025G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 53798-96-2
Molecular Formula C8H12O7
Molecular Weight (g/mol) 220.18
Synonym 2-Hydroxy-1,2,3-propanetricarboxylic Acid 1,3-Dimethyl Ester, sym-Dimethyl Citrate, 2-Hydroxy-4-methoxy-2-(2-methoxy-2-oxoethyl)-4-oxobutanoic Acid, α,α'-Dimethyl Citrate
SMILES COC(=O)CC(CC(=O)OC)(C(=O)O)O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C8H12O7/c1-14-5(9)3-8(13,7(11)12)4-6(10)15-2/h13H,3-4H2,1-2H3,(H,11,12)/p-1
Formula Weight 220.0583
Chemical Name or Material 1,5-Dimethyl Citrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.