missing translation for 'onlineSavingsMsg'
Learn More

(1,5-Dimethyl-1H-pyrrol-2-yl)methylamine, 95+%, Thermo Scientific™

Product Code. 10099561
Click to view available options
Quantity:
250 mg
1 g
10 g
Unit Size:
10g
1g
250mg
This item is not returnable. View return policy

Product Code. 10099561

Brand: Thermo Scientific Maybridge SP01447CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 118799-24-9
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.187
InChI Key KMUHAJOXPNPERW-UHFFFAOYSA-N
Synonym 1,5-dimethyl-1h-pyrrol-2-yl methylamine, 1,5-dimethyl-1h-pyrrol-2-yl methanamine, 1,5-dimethylpyrrol-2-yl methanamine, 1,5-dimethyl-1h-pyrrol-2-yl methyl amine, 1-1,5-dimethyl-1h-pyrrol-2-yl methanamine, 1,5-dimethylpyrrol-2-yl methylamine, 1-1,5-dimethylpyrrol-2-yl methanamine, 1h-pyrrole-2-methanamine, 1,5-dimethyl, 2-aminomethyl-1,5-dimethyl-1h-pyrrole, 1h-pyrrole-2-methanamine, 1,5-dimethyl-9ci
PubChem CID 737232
IUPAC Name (1,5-dimethylpyrrol-2-yl)methanamine
SMILES CC1=CC=C(N1C)CN

Specifications

Assay Percent Range 90%
Quantity 250 mg
Formula Weight 124.19
Percent Purity 90%
Chemical Name or Material (1, 5-Dimethyl-1H-pyrrol-2-yl)methylamine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.