missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dimethyl-1H-pyrazole-3-carboxylic acid, 95+%, Thermo Scientific™

Product Code. 10178264
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10178264 25 g 25g
10364322 1 g 1g
10442064 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10178264 Supplier Thermo Scientific Maybridge Supplier No. CC12601EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5744-59-2
Molecular Formula C6H8N2O2
Molecular Weight (g/mol) 140.14
MDL Number MFCD00085040
InChI Key PXRXGHUTKHXUGF-UHFFFAOYSA-N
Synonym 1,5-dimethyl-1h-pyrazole-3-carboxylic acid, 1,5-dimethyl-1h-pyrazole-3-carboxylicacid, 1,5-dimethyl-3-pyrazolecarboxylic acid, 1h-pyrazole-3-carboxylic acid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic, acmc-209lyb, pyrazole-3-carboxylic acid, 1,5-dimethyl, 1h-pyrazole-3-carboxylicacid, 1,5-dimethyl, 1,5-dimethyl-1h-pyrazole-3-carboxylic acid #
PubChem CID 587757
SMILES CN1N=C(C=C1C)C(O)=O

Specifications

Assay Percent Range 95%
Quantity 25 g
IUPAC Name 1,5-dimethyl-1H-pyrazole-3-carboxylic acid
Formula Weight 140.14
Percent Purity 95%
Chemical Name or Material 1, 5-Dimethyl-1H-pyrazole-3-carboxylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.