missing translation for 'onlineSavingsMsg'
Learn More

1,5-Diiodopentane, 97%, stab. with copper

Product Code. 11453537
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11453537

Brand: Thermo Scientific Alfa Aesar A17298.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 628-77-3 | C5H10I2 | 323.944 g/mol

1,5-Diiodopentane is used as a crosslinking reagent. It is used in the preparation of (iodoalkylaminocarbene)tungsten complexes. It is also used as alkylation to form unsubstituted cyclohexane. It is involved in the preparation of poly(1-vinyl imidazole-co-elthylene glycol methyl ether acrylate electrolytes and used for quasi-solid state dye-sensitized solar cells.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,5-Diiodopentane is used as a crosslinking reagent. It is used in the preparation of (iodoalkylaminocarbene)tungsten complexes. It is also used as alkylation to form unsubstituted cyclohexane. It is involved in the preparation of poly(1-vinyl imidazole-co-elthylene glycol methyl ether acrylate electrolytes and used for quasi-solid state dye-sensitized solar cells.

Solubility
Immiscible with water.

Notes
Light sensitive. Incompatible with strong oxidizing agents, strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-77-3
Molecular Formula C5H10I2
Molecular Weight (g/mol) 323.944
MDL Number MFCD00001101
InChI Key IAEOYUUPFYJXHN-UHFFFAOYSA-N
Synonym pentane, 1,5-diiodo, pentamethylene diiodide, 1,5-diiodopentan, pentane,5-diiodo, pubchem3853, acmc-20ap1i, 4-01-00-00317 beilstein handbook reference, iaeoyuupfyjxhn-uhfffaoysa, 1,5-diiodopentane, stab. with copper, 1,5-diiodopentane, contains copper as stabilizer
PubChem CID 12354
IUPAC Name 1,5-diiodopentane
SMILES C(CCI)CCI

Specifications

Density 2.176
Boiling Point 128°C to 130°C (10 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.6012
Quantity 25 g
Beilstein 1697281
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 323.94
Percent Purity 97%
Chemical Name or Material 1,5-Diiodopentane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.