missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dihydroxynaphthalene-d6, TRC

Product Code. 30369006
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30369006 100 mg -
30382906 10 mg -
30375767 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369006 Supplier Toronto Research Chemicals Supplier No. TRCD453491100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C10 D6 H2 O2
Molecular Weight (g/mol) 166.206
Synonym 1,5-Dihydroxynaphthaline-d6, 1,5-Naphthohydroquinone-d6
IUPAC Name 2,3,4,6,7,8-hexadeuterionaphthalene-1,5-diol
SMILES [2H]c1c([2H])c(O)c2c([2H])c([2H])c([2H])c(O)c2c1[2H]

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C10H8O2/c11-9-5-1-3-7-8(9)4-2-6-10(7)12/h1-6,11-12H/i1D,2D,3D,4D,5D,6D
Formula Weight 166.09
Chemical Name or Material 1,5-Dihydroxynaphthalene-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.