missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dihydroxyanthraquinone, tech. 90%, Thermo Scientific™

Product Code. 11324324
Change view
Click to view available options
Quantity:
10 g
100 g
500 g
Unit Size:
100g
10g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11324324 500 g 500g
11304324 10 g 10g
11314324 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11324324 Supplier Thermo Scientific Alfa Aesar Supplier No. L00848.36

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 117-12-4
Molecular Formula C14H8O4
Molecular Weight (g/mol) 240.21
MDL Number MFCD00001210
InChI Key JPICKYUTICNNNJ-UHFFFAOYSA-N
Synonym anthrarufin, 1,5-dihydroxyanthraquinone, 1,5-dihydroxyanthrachinon, 9,10-anthracenedione, 1,5-dihydroxy, 1,5-dihydroxy-9,10-anthraquinone, 1,5-dihydroxy-9,10-anthracenedione, anthraquinone, 1,5-dihydroxy, az-u, unii-kpb60w5s3m, ccris 3150
PubChem CID 8328
ChEBI CHEBI:37501
IUPAC Name 1,5-dihydroxyanthracene-9,10-dione
SMILES OC1=CC=CC2=C1C(=O)C1=CC=CC(O)=C1C2=O

Specifications

Melting Point ∽280°C (decomposition)
Quantity 500 g
Beilstein 1881718
Merck Index 14,689
Formula Weight 240.21
Percent Purity 90%
Grade Technical
Chemical Name or Material 1,5-Dihydroxyanthraquinone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.