missing translation for 'onlineSavingsMsg'
Learn More

1,5-Dihydroxy Canagliflozin, TRC

Product Code. 30383990
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30383990 10 mg 10mg
30373389 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30383990 Supplier Toronto Research Chemicals Supplier No. TRCD45043110MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,5-Dihydroxy Canagliflozin
Analyte or Component Names 1,5-Dihydroxy Canagliflozin
Shipping Condition Room Temperature
Molecular Formula C24 H27 F O6 S
Synonym Canagliflozin Enantiomer Ring Opening Impurity, (2S,3R,4R,5R)-1-(3-((5-(4-fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)hexane-1,2,3,4,5,6-hexaol
InChI Formula InChI=1S/C24H27FO6S/c1-13-2-3-15(21(28)23(30)24(31)22(29)19(27)12-26)10-16(13)11-18-8-9-20(32-18)14-4-6-17(25)7-5-14/h2-10,19,21-24,26-31H,11-12H2,1H3/t19-,21?,22-,23+,24+/m1/s1
Solution Type Neat
SMILES Cc1ccc(cc1Cc2ccc(s2)c3ccc(F)cc3)C(O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO
IUPAC Name (2S,3R,4R,5R)-1-[3-[[5-(4-fluorophenyl)thiophen-2-yl]methyl]-4-methylphenyl]hexane-1,2,3,4,5,6-hexol
Industry Type Pharmaceutical
Molecular Weight (g/mol) 462.531
Quantity 10 mg
Formula Weight 462.151
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.