missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dihydroxy Canagliflozin, TRC
Specifications
Specifications
| Chemical Name or Material | 1,5-Dihydroxy Canagliflozin |
| Analyte or Component Names | 1,5-Dihydroxy Canagliflozin |
| Shipping Condition | Room Temperature |
| Molecular Formula | C24 H27 F O6 S |
| Synonym | Canagliflozin Enantiomer Ring Opening Impurity, (2S,3R,4R,5R)-1-(3-((5-(4-fluorophenyl)thiophen-2-yl)methyl)-4-methylphenyl)hexane-1,2,3,4,5,6-hexaol |
| InChI Formula | InChI=1S/C24H27FO6S/c1-13-2-3-15(21(28)23(30)24(31)22(29)19(27)12-26)10-16(13)11-18-8-9-20(32-18)14-4-6-17(25)7-5-14/h2-10,19,21-24,26-31H,11-12H2,1H3/t19-,21?,22-,23+,24+/m1/s1 |
| Solution Type | Neat |
| SMILES | Cc1ccc(cc1Cc2ccc(s2)c3ccc(F)cc3)C(O)[C@H](O)[C@@H](O)[C@H](O)[C@H](O)CO |
| IUPAC Name | (2S,3R,4R,5R)-1-[3-[[5-(4-fluorophenyl)thiophen-2-yl]methyl]-4-methylphenyl]hexane-1,2,3,4,5,6-hexol |
| Industry Type | Pharmaceutical |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?