missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,5-Dichloropentane, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | 1,5-Dichloropentane |
| CAS | 628-76-2 |
| Molecular Formula | C5 H10 Cl2 |
| Quantity | 25 g |
| Synonym | NSC 6210, Pentamethylene Chloride |
| InChI Formula | InChI=1S/C5H10Cl2/c6-4-2-1-3-5-7/h1-5H2 |
| SMILES | ClCCCCCCl |
| IUPAC Name | 1,5-dichloropentane |
| Molecular Weight (g/mol) | 141.039 |
| Formula Weight | 140.016 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction